Merck
  • HIF-prolyl hydroxylase is a potential molecular target for esculetin-mediated anti-colitic effects.
HIF-prolyl hydroxylase is a potential molecular target for esculetin-mediated anti-colitic effects.
Fitoterapia (2015-03-24)
Soohwan Yum, Seongkeun Jeong, Sunyoung Lee, Wooseong Kim, Joon Nam, Yunjin Jung
ABSTRACT
We investigated a potential molecular target for anti-colitic effects of esculetin, 6,7-dihydroxycoumarin. Esculetin administered rectally effectively ameliorated TNBS-induced rat colitis and attenuated the expression of pro-inflammatory mediators in the inflamed colon. In human colon carcinoma HCT116 cells, esculetin induced hypoxia-inducible factor-1α (HIF-1α), leading to secretion of vascular endothelial growth factor, a HIF-1 target gene product involved in ulcer healing of the gastrointestinal mucosa. Esculetin directly inhibited HIF prolyl hydroxylase-2 (HPH-2), an enzyme playing a major role in negatively regulating HIF-1α protein stability. Esculetin inhibition of HPH and consequent induction of HIF-1α were attenuated by escalating dose of either ascorbate or 2-ketoglutarate, the required factors of the enzyme. Structurally, the catechol moiety in esculetin was required for HPH inhibition. Collectively, HPH may be a molecular target for esculetin-mediated anti-colitic effects and the catechol moiety in esculetin is the pharmacophore for HPH inhibition.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
Pyrocatechol, ≥99%
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Sigma-Aldrich
Coumarin, ≥99% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Iron(II) chloride, 98%
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Picrylsulfonic acid solution, 5 % (w/v) in H2O, BioReagent, suitable for determination of primary amines