Merck
  • Hepatitis C virus core protein overcomes H2O2-induced apoptosis by downregulating p14 expression via DNA methylation.
Hepatitis C virus core protein overcomes H2O2-induced apoptosis by downregulating p14 expression via DNA methylation.
The Journal of general virology (2014-12-24)
Young Lan Seo, Shinhee Heo, Kyung Lib Jang
ABSTRACT
Infection with hepatitis C virus (HCV) is characterized by systemic oxidative stress that is caused by either viral core protein or chronic inflammation. It is well recognized that reactive oxygen species (ROS) such as H2O2 can induce apoptotic cell death and can therefore function as anti-tumorigenic species. However, the detailed mechanisms by which ROS induce apoptotic cell death and HCV copes with the oxidative conditions are largely unknown. In the present study, we found that H2O2 induced apoptotic cell death in p53-positive human hepatocytes, but not in p53-negative human hepatocytes. For this effect, H2O2 upregulated levels of p14, increased ubiquitin-dependent degradation of mouse double minute 2 (MDM2), and reduced the interaction between MDM2 and p53 to prevent p53 degradation, resulting in accumulation of p53 and subsequent activation of p53-dependent apoptotic pathways. Interestingly, HCV core repressed p14 expression via promoter hypermethylation to abolish the potential of H2O2 to activate the p14-MDM2-p53 pathway. As a consequence, HCV core-expressing cells could overcome p53-mediated apoptosis provoked by H2O2. Taken together, HCV core could contribute to hepatocellular carcinoma formation by removing deleterious roles of ROS inducing cell death.
Sigma-Aldrich
MISSION® esiRNA, targeting human FABP1
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
MISSION® esiRNA, targeting human S100A9
Sigma-Aldrich
MISSION® esiRNA, targeting human TP53
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydroquinone, meets USP testing specifications
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human SUB1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cdk2ap2
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Sub1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Robld3
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O