Merck
Deferasirox-induced cytogenetic responses.
Environmental toxicology and pharmacology (2015-03-04)
Mehmet Arslan, Hasan Basri Ila
ABSTRACT
Deferasirox (commercially formulated as Exjade(®)) is one of the effective iron chelators used in treatment of iron overload diseases. In this study the effect of this substance for chromosome aberration, sister chromatid exchange and mitotic index was studied by in vitro (by using human peripheral lymphocytes) and in vivo (by using rat) analysis. Deferasirox increased the sister chromatid exchange frequency in all tested concentrations and periods in vitro. Also, in the presence of metabolic activator, the substance led to a statistically significant increase in the sister chromatid exchange frequencies only at high concentration. While in in vitro analysis the substance significantly increased abnormal cell percentages in all concentrations, in in vivo study the substance increased chromosome aberrations only in two concentrations at 12h treatment. In the cultured lymphocytes, deferasirox showed cytotoxicity by significantly reducing proliferation index and mitotic index values. While in the presence of metabolic activation it did not affect the proliferation index frequency, it had a stimulant effect on the mitotic index frequency. Deferasirox reduced significantly the mitotic index value in the bone marrow cells especially in high concentration and short treatment period (12h).
Sigma-Aldrich
Magnesium methyl carbonate solution, 2.0 M in DMF
Sigma-Aldrich
Urethane, ≥99.0% (GC)
Sigma-Aldrich
Colchicine, BioReagent, suitable for plant cell culture, ≥95% (HPLC)
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder
Sigma-Aldrich
Urethane, ≥99%