Merck
  • Synergy and Other Interactions between Polymethoxyflavones from Citrus Byproducts.
Synergy and Other Interactions between Polymethoxyflavones from Citrus Byproducts.
Molecules (Basel, Switzerland) (2015-11-13)
Benito F García, Ascensión Torres, Francisco A Macías
ABSTRACT
The citrus by-products released from citrus processing plants may contain high levels of potentially bioactive compounds such as flavonoids, which are a widely distributed group of polyphenolic compounds with health-related properties based on their antioxidant activity. In the study reported here, the potential bioactivities and antioxidant activities of extracts, fractions and compounds from citrus by-products were evaluated along with the chemical interactions of binary mixtures of compounds and complex mixtures. The bioactivities and interactions were evaluated in wheat coleoptile bioassays and the antioxidant activity was evaluated by the al DPPH (2,2-diphenyl-1-picrylhdrazyl radical) radical scavenging assay. The extracts, fractions and most of the isolated compounds (mainly polymethoxyflavones) showed high activity in the wheat coleoptile bioassay. However, the antioxidant activity was not consistently high, except in the acetone extract fractions. Moreover, a study of the interactions with binary mixtures of polymethoxyflavones showed the occurrence of synergistic effects. The complex mixtures of fractions composed mainly of polymethoxyflavones caused a synergistic effect when it was added to a bioactive compound such as anethole. The results reported here highlight a new application for the wheat coleoptile bioassay as a quick tool to detect potential synergistic effects in compounds or mixtures.
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%