Merck
  • Determination of 2-Methylimidazole, 4-Methylimidazole, and 2-Acetyl-4-(1,2,3,4-tetrahydroxybutyl)imidazole in Licorice Using High-Performance Liquid Chromatography-Tandem Mass Spectrometry Stable-Isotope Dilution Analysis.
Determination of 2-Methylimidazole, 4-Methylimidazole, and 2-Acetyl-4-(1,2,3,4-tetrahydroxybutyl)imidazole in Licorice Using High-Performance Liquid Chromatography-Tandem Mass Spectrometry Stable-Isotope Dilution Analysis.
Journal of agricultural and food chemistry (2015-06-16)
Marion Raters, Paul W Elsinghorst, Stephanie Goetze, Anna Dingel, Reinhard Matissek
ABSTRACT
A quick and selective analytical method was developed for the simultaneous quantitation of 2-methylimidazole, 4-methylimidazole, and 2-acetyl-4-(1,2,3,4-tetrahydroxybutyl)imidazole, which are known to be formed by Maillard reactions. The methodology reported here employs stable-isotope dilution analysis (SIDA) using 4-methylimidazole-d6 and [(13)C6]-2-acetyl-4-(1,2,3,4-tetrahydroxybutyl)imidazole as internal standards. It was successfully applied in a model assay to show that the addition of ammonium chloride during the manufacture of licorice promotes imidazole formation depending on the added amount of ammonium chloride without the well-known impact of present caramel food colorings. Furthermore, a monitoring assay of 29 caramel coloring-free licorice products showed that both 4-methylimidazole and 2-acetyl-4-(1,2,3,4-tetrahydroxybutyl)imidazole are endogenously generated in detectable quantities. None of the samples showed 2-methylimidazole levels above the limit of detection, 50 μg/kg.
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Potassium hexacyanoferrate(II) trihydrate, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
LDN-57444, ≥98% (HPLC)
Sigma-Aldrich
Potassium hexacyanoferrate(II) trihydrate, ≥99.95% trace metals basis
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.