Merck
  • The p53 family member p73 modulates the proproliferative role of IGFBP3 in short children born small for gestational age.
The p53 family member p73 modulates the proproliferative role of IGFBP3 in short children born small for gestational age.
Molecular biology of the cell (2015-06-13)
Flaviana Marzano, Annamaria Ventura, Mariano Francesco Caratozzolo, Italia Aiello, Francesca Mastropasqua, Giacomina Brunetti, Luciano Cavallo, Elisabetta Sbisà, Maria Felicia Faienza, Apollonia Tullo
ABSTRACT
The regulation of insulin-like growth factor-binding protein 3 (IGFBP3) gene expression is complex, because it can be induced by agents that both stimulate and inhibit the proliferation. The principal aim of this study was to investigate whether p73, a member of the p53 gene family, has a role in the regulation of the IGFBP3 expression and whether this regulation occurs in a context of cell survival or death. We demonstrate that IGFBP3 is a direct TAp73α (the p73 isoform that contains the trans-activation domain) target gene and activates the expression of IGFBP3 in actively proliferating cells. As IGFBP3 plays a key role in regulating the growth hormone/insulin-like growth factor type 1 (GH/IGF1) axis, whose alterations in gene expression appear to have a role in the growth failure of children born small for gestational age (SGA), we measured the mRNA expression levels of p73 and IGFBP3 in a group of SGA children. We found that mRNA expression levels of p73 and IGFBP3 are significantly lower in SGA children compared with controls and, in particular, p73 mRNA expression is significantly lower in SGA children with respect to height. Our results shed light on the intricate GH/IGF pathway, suggesting p73 as a good biomarker of the clinical risk for SGA children to remain short in adulthood.
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Arhgap24
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
MISSION® esiRNA, targeting human ARHGAP24
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Trp73
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)