Merck
  • Prothrombin activator-like toxin appears to mediate cardiovascular collapse following envenoming by Pseudonaja textilis.
Prothrombin activator-like toxin appears to mediate cardiovascular collapse following envenoming by Pseudonaja textilis.
Toxicon : official journal of the International Society on Toxinology (2015-05-12)
Janeyuth Chaisakul, Geoffrey K Isbister, Margaret A O'Leary, Helena C Parkington, A Ian Smith, Wayne C Hodgson, Sanjaya Kuruppu
ABSTRACT
Brown snake (Pseudonaja spp.)-induced early cardiovascular collapse is a life-threatening medical emergency in Australia. We have previously shown that this effect can be mimicked in animals and is mediated via the release of endogenous mediators. In the present study, we aimed to purify and characterize the component in Pseudonaja textilis venom which induces cardiovascular collapse following envenoming. The component (fraction 3) was isolated using a combination of techniques including hydroxyapatite and reverse phase chromatography. Fraction 3 (10 or 20 μg/kg, i.v.) produced a rapid decrease in mean arterial pressure (MAP) followed by cardiovascular collapse. Fraction 3-induced early collapse was abolished by prior administration of smaller priming doses of fraction 3 (i.e. 2 and 5 μg/kg, i.v.) or heparin (300 units/kg, i.v.). P. textilis whole venom (1 and 3 μg/ml), but not fraction 3 (1 or 3 μg/ml), induced endothelium-dependent relaxation in isolated rat mesenteric arteries. SDS-PAGE gel indicated the presence of 9-10 protein bands of fraction 3. Using proteomic based analysis some protein bands of fraction 3 were identified as subunits of venom prothrombin activator, pseutarin C of P. textilis venom. Our results conclude that prothrombin activator-like toxin is likely to be a contributor to the rapid collapse induced by P. textilis venom.
Sigma-Aldrich
Hydroxyapatite, purum p.a., ≥90% (as Ca3(PO4)2, KT)
Sigma-Aldrich
Hydroxyapatite, reagent grade, powder, synthetic
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), ≥97%, synthetic
Sigma-Aldrich
Calcium phosphate tribasic, suitable for plant cell culture, BioReagent, powder
Sigma-Aldrich
Calcium phosphate tribasic, 34.0-40.0% Ca basis
Sigma-Aldrich
Acetylcholine bromide, ≥99%
Sigma-Aldrich
Hydroxyapatite, synthetic, 99.8% trace metals basis (excludes Mg)
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), contains 5 wt. % silica as dopant, synthetic
Sigma-Aldrich
Hydroxyapatite, puriss., meets analytical specification of Ph. Eur., BP, FCC, E341, ≥90% (calculated on glowed substance)
Sigma-Aldrich
Hydroxyapatite, nanoparticles, dispersion, 10 wt. % in H2O, <200 nm particle size (BET)
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
3-[(1R)-1-Hydroxy-2-(methylamino)ethyl]phenol, AldrichCPR
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Pyrilamine maleate salt
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ultrapure Acetonitrile