Merck
  • Antioxidant and Anti-inflammatory Activities of Essential Oil and Extracts of Piper miniatum.
Antioxidant and Anti-inflammatory Activities of Essential Oil and Extracts of Piper miniatum.
Natural product communications (2016-01-12)
Wan Mohd Nuzul Hakimi Salleh, Mohd Fariz Kammil, Farediah Ahmad, Hasnah Mohd Sirat
ABSTRACT
The chemical composition of the essential oil and antioxidant and anti-inflammatory activities of the extracts from Piper miniatum were determined. GC and GC-MS analysis of the essential oil resulted in the identification of 64 components, accounting for 89.2% of the total. The major components were caryophyllene oxide (20.3%) and α-cubebene (10.4%). The antioxidant activity was evaluated by β-carotene/linoleic acid bleaching, DPPH radical scavenging and total phenolic content. In the β-carotene assay, the n-hexane extract showed the highest inhibition activity with 42.7%, while the oil gave 91.3%. The essential oil and extracts were tested for anti-inflammatory activity by using the TPA-induced mouse ear edema model and lipoxygenase assays. The essential oil exhibited significant activity in both models as an anti-inflammatory agent. The n-hexane extract showed strong activity with inhibition of 85.9% in the TPA-induced mouse ear edema model, while the chloroform extract showed the highest activity with 94.2% in the lipoxygenase assay.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
1-Phenyl-3-pyrazolidinone, 97%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Linoleic acid, ≥99%
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%