Merck
  • Tl(I) and Tl(III) alter the expression of EGF-dependent signals and cyclins required for pheochromocytoma (PC12) cell-cycle resumption and progression.
Tl(I) and Tl(III) alter the expression of EGF-dependent signals and cyclins required for pheochromocytoma (PC12) cell-cycle resumption and progression.
Journal of applied toxicology : JAT (2014-12-24)
María T L Pino, Sandra V Verstraeten
ABSTRACT
The effects of thallium [Tl(I) and Tl(III)] on the PC12 cell cycle were evaluated without (EGF(-)) or with (EGF(+)) media supplementation with epidermal growth factor (EGF). The following markers of cell-cycle phases were analyzed: cyclin D1 (G1 ); E2F-1, cyclin E and cytosolic p21 (G1 →S transition); nuclear PCNA and cyclin A (S); and cyclin B1 (G2). The amount of cells in each phase and the activation of the signaling cascade triggered by EGF were also analyzed. Tl(I) and Tl(III) (5-100 μM) caused dissimilar effects on PC12 cell proliferation. In EGF(-) cells, Tl(I) increased the expression of G1 →S transition markers and nuclear PCNA, without affecting cyclin A or cyclin B1. In addition to those, cyclin B1 was also increased in EGF(+) cells. In EGF(-) cells, Tl(III) increased the expression of cyclin D1, all the G1→S and S phase markers and cyclin B1. In EGF(+) cells, Tl(III) increased cyclin D1 expression and decreased all the markers of G1 →S transition and the S phase. Even when these cations did not induce the activation of EGF receptor (EGFR) in EGF(-) cells, they promoted the phosphorylation of ERK1/2 and Akt. In the presence of EGF, the cations anticipated EGFR phosphorylation without affecting the kinetics of EGF-dependent ERK1/2 and Akt phosphorylation. Altogether, results indicate that Tl(I) promoted cell proliferation in both EGF(-) and EGF(+) cells. In contrast, Tl(III) promoted the proliferation of EGF(-) cells but delayed it in EGF(+) cells, which may be related to the toxic effects of this cation in PC12 cells.
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Thallium(I) nitrate, 99.9% trace metals basis
Sigma-Aldrich
Thallium(I) nitrate, 99.999% trace metals basis
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)