Merck
  • The mitochondrial ubiquitin ligase MARCH5 resolves MAVS aggregates during antiviral signalling.
The mitochondrial ubiquitin ligase MARCH5 resolves MAVS aggregates during antiviral signalling.
Nature communications (2015-08-08)
Young-Suk Yoo, Yong-Yea Park, Jae-Hoon Kim, Hyeseon Cho, Song-Hee Kim, Ho-Soo Lee, Tae-Hwan Kim, You Sun Kim, Youngsoo Lee, Chul-Joong Kim, Jae U Jung, Jong-Soo Lee, Hyeseong Cho
ABSTRACT
Mitochondria serve as platforms for innate immunity. The mitochondrial antiviral signalling (MAVS) protein forms aggregates that elicit robust type-I interferon induction on viral infection, but persistent MAVS signalling leads to host immunopathology; it remains unknown how these signalling aggregates are resolved. Here we identify the mitochondria-resident E3 ligase, MARCH5, as a negative regulator of MAVS aggregates. March5(+/-) mice and MARCH5-deficient immune cells exhibit low viral replication and elevated type-I interferon responses to RNA viruses. MARCH5 binds MAVS only during viral stimulation when MAVS forms aggregates, and these interactions require the RING domain of MARCH5 and the CARD domain of MAVS. MARCH5, but not its RING mutant (MARCH5(H43W)), reduces the level of MAVS aggregates. MARCH5 transfers ubiquitin to Lys7 and Lys500 of MAVS and promotes its proteasome-mediated degradation. Our results indicate that MARCH5 modulates MAVS-mediated antiviral signalling, preventing excessive immune reactions.
Sigma-Aldrich
MISSION® esiRNA, targeting human MARCH5
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting mouse March5
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%