Merck
  • Platinum-Incorporating Poly(N-vinylpyrrolidone)-poly(aspartic acid) Pseudoblock Copolymer Nanoparticles for Drug Delivery.
Platinum-Incorporating Poly(N-vinylpyrrolidone)-poly(aspartic acid) Pseudoblock Copolymer Nanoparticles for Drug Delivery.
Biomacromolecules (2015-05-30)
Xikuang Yao, Chen Xie, Weizhi Chen, Chenchen Yang, Wei Wu, Xiqun Jiang
ABSTRACT
Cisplatin-incorporating pseudoblock copolymer nanoparticles with high drug loading efficiency (ca. 50%) were prepared built on host-guest inclusion complexation between β-cyclodextrin end-capped poly(N-vinylpyrrolidone) block and admantyl end-capped poly(aspartic acid) block, followed by the coordination between cisplatin and carboxyl groups in poly(aspartic acid). The host-guest interaction between the two polymer blocks was examined by two-dimensional nuclear overhauser effect spectroscopy. The size and morphology of nanoparticles formed were characterized by dynamic light scattering, zeta potential, transmission electron microscopy, and atomic force microscopy. The size control of nanoparticles was carried out by varying the ratio of poly(N-vinylpyrrolidone) to poly(aspartic acid). The nanoparticles were stable in the aqueous medium with different pH values but disintegrated in the medium containing Cl(-) ions. The in vitro and in vivo antitumor effects of cisplatin-loaded nanoparticles were evaluated. The biodistribution of the nanoparticles in vivo was studied by noninvasive near-infrared fluorescence imaging and ion-coupled plasma mass spectrometry. It was found that cisplatin-loaded nanoparticles could effectively accumulate in the tumor site and exhibited significant superior in vivo antitumor activity to the commercially available free cisplatin by combining the tumor volume, body weight, and survival rate measurements.
Sigma-Aldrich
Propargyl alcohol, 99%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
SAFC
L-Aspartic acid
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Propionic acid, BioReagent, suitable for insect cell culture, ~99%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Amantadine hydrochloride
Sigma-Aldrich
Propionic acid, natural, 99%, FG
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Propionic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Propionic acid, puriss. p.a., ≥99.5% (GC)