Merck
Effect of n-alkanes on lipid bilayers depending on headgroups.
Chemistry and physics of lipids (2015-05-11)
Mafumi Hishida, Asami Endo, Koyomi Nakazawa, Yasuhisa Yamamura, Kazuya Saito
ABSTRACT
Phase behavior and structural properties were examined for phospholipid bilayers having different headgroups (DMPC, DMPS and DMPE) with added n-alkanes to study effect of flexible additives. Change in the temperatures of main transition of the lipid/alkane mixtures against the length of added alkanes depends largely on the headgroup. Theoretical analysis of the change of the temperature of transition indicates that the headgroup dependence is dominantly originated in the strong dependence of total enthalpy on the headgroups. The results of X-ray diffraction show that the enthalpic stabilization due to enhanced packing of acyl chains of the lipid by alkanes in the gel phase causes the headgroup-dependent change in the phase transition behavior. The enhanced packing in the gel phase also leads to easy emergence of the subgel phase with very short relaxation time at room temperature in the DMPE-based bilayers.
Sigma-Aldrich
Decane, anhydrous, ≥99%
Sigma-Aldrich
Decane, ≥95%
Sigma-Aldrich
Octane, anhydrous, ≥99%
Sigma-Aldrich
Octane, electronic grade, ≥99.999% metals basis, ≥99% (CP)
Sigma-Aldrich
Dodecane, anhydrous, ≥99%
Sigma-Aldrich
Tetradecane, ≥99%
Sigma-Aldrich
Tetradecane, olefin free, ≥99.0% (GC)
Sigma-Aldrich
Sodium 2,3-dimercaptopropanesulfonate monohydrate, 95%