Merck
  • Hypomethylated Rab27b is a progression-associated prognostic biomarker of glioma regulating MMP-9 to promote invasion.
Hypomethylated Rab27b is a progression-associated prognostic biomarker of glioma regulating MMP-9 to promote invasion.
Oncology reports (2015-07-15)
Hongjun Wang, Yongzhi Wang, Zhaoshi Bao, Chuanbao Zhang, Yanwei Liu, Jinquan Cai, Chuanlu Jiang
ABSTRACT
In the present study, in order to analyze abnormalities in DNA methylation in glioma, we utilized a large cohort methylation microarray (119 glioma samples). Genes associated with tumor grade progression were screened through Significance Analysis of Microarrays (SAM) in the methylation microarray. We found that Rab27b was hypomethylated in high-grade glioma (anaplastic gliomas and glioblastomas) compared with low-grade glioma (astrocytoma, oligodendrocytoma and oligoastrocytoma) (p=0.02). In 52 glioma samples, we determined both the methylation status of the Rab27b promoter region and protein expression, and confirmed a negative correlation between the methylation status and expression (p<0.01). Immunohistochemistry of 91 gliomas revealed that the Rab27b expression scores of high-grade glioma were higher than scores of low-grade gliomas (p<0.01). In high‑grade gliomas, patients haboring Rab27b hypomethylation or overexpression had unfavorable survival prognosis. Transwell invasion assays identified that invasive cell number of glioma U87 and LN229 cells decreased when Rab27b was knocked down. Decreased invasion partly resulted from reduced expression and activation of MMP-9 after Rab27b knockdown. Downregulation of Rab27b also suppressed tumor growth in vivo. Hypomethylated Rab27b was identified as a progression-associated and prognostic molecular marker of glioma.
Sigma-Aldrich
MISSION® esiRNA, targeting human RAB27B (2)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Rab27b
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri