Merck
Hesperetin prevents selenite-induced cataract in rats.
Molecular vision (2015-08-19)
Yosuke Nakazawa, Mikako Oka, Masayasu Bando, Makoto Takehana
ABSTRACT
This study investigated the ability of hesperetin, a natural flavonoid, to prevent selenite-induced cataracts in a rat model. Animals were divided into four treatment groups: G1 (control group), G2 (hesperetin-treated group), G3 (selenite-induced cataract group), and G4 (hesperetin-treated selenite cataract group). Animals in the G1 and G3 groups were injected with vehicle alone, while those in the G2 and G4 groups received a subcutaneous injection of hesperetin (0.4 μg/g bodyweight on days 0, 1, and 2, corresponding to P13, P14, and P15). Sodium selenite (20 μmol/g bodyweight given 4 h after the hesperetin injection on day 0) was administered to rats in the G3 and G4 groups to induce cataract formation. Lenses were observed with slit-lamp microscopy, and filensin degradation and the decreased glutathione (GSH) and ascorbic acid levels in the lens were measured on day 6. Lenses in the G3 group showed mature central opacity, while some lenses in the G4 group lacked central opacity and had lower-grade cataracts. All lenses in the G1 and G2 groups were transparent. Expression of the 94 kDa and 50 kDa forms of filensin was significantly decreased in the lenses in the G3 group compared with those in the G1 and G2 groups. Interestingly, these forms of filensin rescued the rat lenses in the G4 group. In the G3 group lenses, the GSH and ascorbic acid levels were lower than in the control group but were normalized in the G4 group lenses. The results suggest that hesperetin can prevent selenite-induced cataract formation.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium selenite, 99%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Hesperetin, ≥95%
Sigma-Aldrich
3,3′-Diaminobenzidine, 97%
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Tropicamide, solid
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
3,3′-Diaminobenzidine, 97% (HPLC)
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder