Merck
  • A novel approach for predicting the dissolution profiles of pharmaceutical tablets.
A novel approach for predicting the dissolution profiles of pharmaceutical tablets.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2015-07-15)
Raphael Paus, Elena Hart, Yuanhui Ji
ABSTRACT
In this paper, the intrinsic dissolution profiles of naproxen (NAP) at pH values of 1.5 and 3.0 and of trimethoprim (TMP) at pH values of 1.5, 3.0, 5.0, 6.5 and 7.2 were measured. Meanwhile, the dissolution profiles of NAP and TMP from cylindrical tablets were measured at different temperatures (298.15K, 305.15K, 301.15K and 310.15K) and stirring speeds (50rpm, 100rpm and 150rpm) as well as at different pH values (1.5, 3.0, 5.0, 6.5 and 7.2). Additionally the pH-dependent solubilities of both APIs were measured and modeled. The chemical-potential-gradient model combined with the perturbed-chain statistical associating fluid theory (PC-SAFT) was applied to predict the dissolution profiles of the cylindrical tablets of NAP and TMP under different conditions based on the analysis of their intrinsic dissolution profiles as well as on the determination of the surface-area reduction of the API tablets during dissolution. It was shown that the predicted dissolution profiles of the tablets under different conditions were in a good accordance with the experimental findings.
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Naproxen, meets USP testing specifications
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Trimethoprim, ≥98.5%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
di-Sodium hydrogen phosphate dihydrate, BioUltra, for molecular biology, ≥99.0% (T)
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
N-Acetyl-D-penicillamine, for HPLC derivatization, ≥99.0% (T)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%