Merck
  • A Highly Specific, Fully Validated Urinary Ethyl Glucuronide Analysis Using Solid Phase Extraction-Liquid Chromatography/Electrospray Ionization-Tandem Mass Spectrometry.
A Highly Specific, Fully Validated Urinary Ethyl Glucuronide Analysis Using Solid Phase Extraction-Liquid Chromatography/Electrospray Ionization-Tandem Mass Spectrometry.
Journal of AOAC International (2015-06-19)
Munevver Acikkol, Ali Acar Ozdemir, Beril Anilanmert, Hasan Mirsal
ABSTRACT
A highly specific and selective analytical method using LC/MS/MS for the quantitative determination of ethyl glucuronide (EtG) in urine was developed and fully validated. Since the determination of EtG in urine may be possible days after the elimination of alcohol, it is an indication of alcohol use in alcohol treatment programs and antemortem and postmortem toxicological investigations. Propyl glucuronide (PrG), which increased the selectivity of the method, was used as an internal standard. The method was validated in terms of selectivity, linearity, LOD, LOQ, intraday and interday precision, and recovery. The analyte and PrG in the SPE cleaned up extract were separated on a 150 mm C18 column in 3.3 min with high resolution. The rest of the peaks from the matrix were eluted in 9.0 min. The LOD was 90.8 ng/mL and the LOQ was calculated using the EURACHEM method as 185.0 ng/mL. The intraday and the intermediate precision of the method was calculated using analysis of variance and confirmed with calculation of HorRat values, which were found within acceptable limits. The method provided a reliable solution for monitoring patients under alcohol addiction treatment and was successfully applied to real samples.
Sigma-Aldrich
Picric acid solution, 1.3% in H2O (saturated)
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
tert-Butanol, anhydrous, ≥99.5%
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Picric acid, moistened with water, ≥98%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%