Merck
  • p85 protein expression is associated with poor survival in HER2-positive patients with advanced breast cancer treated with trastuzumab.
p85 protein expression is associated with poor survival in HER2-positive patients with advanced breast cancer treated with trastuzumab.
Pathology oncology research : POR (2014-08-08)
Kitty Pavlakis, Mattheos Bobos, Anna Batistatou, Vassiliki Kotoula, Anastasia G Eleftheraki, Anastasios Stofas, Eleni Timotheadou, George Pentheroudakis, Amanda Psyrri, Angelos Koutras, Dimitrios Pectasides, Pavlos Papakostas, Evangelia Razis, Christos Christodoulou, Konstantine T Kalogeras, George Fountzilas
ABSTRACT
To investigate the immunohistochemical expression of p85 in a cohort of trastuzumab-treated HER2-positive and HER2-negative metastatic breast cancer patients. The medical records of all patients with metastatic breast cancer treated with trastuzumab-based regimens between 1998 and 2010 were reviewed and clinical information was obtained. Formalin-fixed paraffin-embedded tumor tissue samples with adequate material were retrospectively collected from 183 patients. Samples were evaluated by immunohistochemistry for p85, estrogen receptors (ER), progesterone receptors (PgR), HER2, Ki67, PTEN and phosphorylated Akt (S473 and T308). HER2 status was studied by fluorescence in situ hybridization, as well. PIK3CA mutational status was also evaluated. Median follow-up for all patients was 72 months. Central re-evaluation for HER2 revealed only 111 HER2-positive cases, with the remaining 72 patients being HER2-negative. Median survival was longer in HER2-positive patients (50.7 months) compared to HER2-negative patients (36.6 months) both treated with trastuzumab, but this difference has not reached significance (p = 0.068). In total, 62% of the patients were found positive for p85, however the p85 protein was not found to be differentially expressed in HER2-positive versus HER2-negative cases. There were no significant associations between protein expression of p85 and any of the markers under study, or with time to progression. Positive p85 protein expression was however associated with poor survival in trastuzumab-treated HER2-positive patients. In our cohort of trastuzumab-treated HER2-positive breast cancer patients, positive p85 protein expression appears to be a prognostic factor of poor survival and, if validated, might have important implications in the treatment of such patients.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Trimethylaluminum, packaged for use in deposition systems
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
SAFC
L-Threonine
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Hydrogen sulfide, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Trimethylaluminum, 97%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Trimesic acid, 95%
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in heptane
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in hexanes
Sigma-Aldrich
Sulfur-32S, 99.9 atom % 32S
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in toluene
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
Hydrogen sulfide solution, 0.8 M in THF