Merck
  • miR-330-3p controls cell proliferation by targeting early growth response 2 in non-small-cell lung cancer.
miR-330-3p controls cell proliferation by targeting early growth response 2 in non-small-cell lung cancer.
Acta biochimica et biophysica Sinica (2015-05-04)
Xuzhi Liu, Hanbing Shi, Bo Liu, Jianing Li, Yaxin Liu, Baiquan Yu
ABSTRACT
Non-small-cell lung cancer (NSCLC) is one of the most common lung cancers, and microRNAs (miRNAs) have been reported to play essential roles in NSCLC. Recent studies have indicated that miR-330-3p expression is up-regulated in NSCLC samples and in tissues of NSCLC brain metastasis. In this study, up-regulation of miR-330-3p expression was confirmed in NSCLC and 20 NSCLC patient samples. Furthermore, miR-330-3p was over-expressed in NSCLC cell lines A549 and H23, and the promotive function of miR-330-3p was investigated in regulating NSCLC cell proliferation and cell cycle distribution. To identify potential target genes of miR-330-3p in NSCLC, the miRNA target prediction databases were used. Luciferase activity assay and real-time RT-PCR analysis confirmed that miR-330-3p is negatively correlated with the expression of early growth response 2 (EGR2). Moreover, it was also found that EGR2 mRNA contains two potential binding sites for miR-330-3p. Knock-down of EGR2 with siRNA was demonstrated to have a similar effect as the over-expression of miR-330-3p in NSCLC cell lines. Taken together, our results show that EGR2 is a target of miR-330-3p.
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Egr2
Sigma-Aldrich
MISSION® esiRNA, targeting human EGR2
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis