Merck
  • Structural characterization of a polysaccharide from Chrysanthemum morifolium flowers and its antioxidant activity.
Structural characterization of a polysaccharide from Chrysanthemum morifolium flowers and its antioxidant activity.
Carbohydrate polymers (2015-06-17)
Changping Zheng, Qun Dong, Huanjun Chen, Qifei Cong, Kan Ding
ABSTRACT
The flowers of Chrysanthemum morifolium were extracted by boiling water, and a water-soluble polysaccharide (CMJA0S2) with a molecular weight of 6.5 kDa was isolated by anion-exchange chromatography on a DEAE-cellulose column and gel permeation chromatography on a Sephacryl S-300 HR column. Monosaccharide composition analysis indicated that CMJA0S2 was composed of galactose, glucose, mannose and arabinose in molar ratio of 4.1: 3.3: 1.0: 2.3. According to linkage analysis, partial acid hydrolysis and NMR spectra, the backbone was shown to contain 1, 4-linked β-Galp, 1, 4-linked β-Glcp and 1, 4-linked β-Manp, with branches substituted at C-6 of 1, 4-linked β-Manp by 1, 6-linked β-Galp and at O-6 of partial 1, 4-linked β-Galp substituted by T-linked α-Glcp. About 40% of 1, 6-linked β-Galp with T-linked α-Glcp was substituted at O-3 by α-Araf-(1→[5)-α-Araf-(1]3. The anti-oxidative analysis showed that CMJA0S2 could scavenge the DPPH radicals and relieve the damage of PC12 cells caused by H2O2, thus it could be regarded as a potential natural antioxidant.
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Iodomethane-12C, ≥99.9 atom % 12C, ≥99% (CP), contains copper as stabilizer
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Iodomethane, purum, ≥99.0% (GC)
Sigma-Aldrich
Iodomethane, contains copper as stabilizer, ReagentPlus®, 99%
Sigma-Aldrich
Iodomethane, contains copper as stabilizer, ReagentPlus®, 99.5%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Iodomethane solution, 2.0 M in tert-butyl methyl ether, contains copper as stabilizer
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%