Merck
  • Employing Tryptone as a General Phase Transfer Agent to Produce Renal Clearable Nanodots for Bioimaging.
Employing Tryptone as a General Phase Transfer Agent to Produce Renal Clearable Nanodots for Bioimaging.
Small (Weinheim an der Bergstrasse, Germany) (2015-04-29)
Fuyao Liu, Xiuxia He, Junping Zhang, Huimao Zhang, Zhenxin Wang
ABSTRACT
Hydrophobic ultrasmall nanoparticles synthesized in nonpolar solvents exhibit great potential in biomedical applications. However, a major challenge when applying these nanomaterials in biomedical research is the lack of a versatile strategy to render them water dispersible while preserving the hydrodynamic diameter (HD) to be less than 8 nm for efficient renal clearance. To address this problem, tryptone is employed as the novel ligand to fabricate a simple, versatile, and inexpensive strategy for transferring hydrophobic NaGdF(4) nanodots (3 nm in diameter) from organic phase into aqueous phase without any complicated organic synthesis. The paramagnetic properties of NaGdF(4) nanodots are well retained after the phase transfer process. In particular, the tryptone-NaGdF(4) nanodots have ultrasmall HD (ca., 7.5 nm), which greatly improves their tumor accumulation and facilitates renal clearance within 24 h postinjection. The as-prepared tryptone-NaGdF(4) nanodots can also be further functionalized with other molecules for extensively biomedical and bioanalytical applications. Furthermore, the proposed strategy can easily be extended to transfer other types of inorganic nanoparticles from hydrophobic to hydrophilic for facilitating biomedical applications.
Sigma-Aldrich
Isoamyl ether, 99%
Sigma-Aldrich
N-Hydroxysulfosuccinimide sodium salt, ≥98% (HPLC)
Sigma-Aldrich
Argon, ≥99.998%
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Sodium oleate, ≥82% (fatty acids), powder
Sigma-Aldrich
1-Octadecene, ≥95.0% (GC)
Sigma-Aldrich
1-Octadecene, technical grade, 90%
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleylamine, technical grade, 70%
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium oleate, ≥95% (capillary GC)
Sigma-Aldrich
Sodium oleate, ≥99%
Sigma-Aldrich
Argon-40Ar, 99.95 atom %
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
SAFC
Oleylamine, ≥98% (primary amine)