Merck
  • Synthesis, Density Functional Theory (DFT), Urease Inhibition and Antimicrobial Activities of 5-Aryl Thiophenes Bearing Sulphonylacetamide Moieties.
Synthesis, Density Functional Theory (DFT), Urease Inhibition and Antimicrobial Activities of 5-Aryl Thiophenes Bearing Sulphonylacetamide Moieties.
Molecules (Basel, Switzerland) (2015-11-12)
Mnaza Noreen, Nasir Rasool, Yasmeen Gull, Muhammad Zubair, Tariq Mahmood, Khurshid Ayub, Faiz-Ul-Hassan Nasim, Asma Yaqoob, Muhammad Zia-Ul-Haq, Vincenzo de Feo
ABSTRACT
A variety of novel 5-aryl thiophenes 4a-g containing sulphonylacetamide (sulfacetamide) groups were synthesized in appreciable yields via Pd[0] Suzuki cross coupling reactions. The structures of these newly synthesized compounds were determined using spectral data and elemental analysis. Density functional theory (DFT) studies were performed using the B3LYP/6-31G (d, p) basis set to gain insight into their structural properties. Frontier molecular orbital (FMOs) analysis of all compounds 4a-g was computed at the same level of theory to get an idea about their kinetic stability. The molecular electrostatic potential (MEP) mapping over the entire stabilized geometries of the molecules indicated the reactive sites. First hyperpolarizability analysis (nonlinear optical response) were simulated at the B3LYP/6-31G (d, p) level of theory as well. The compounds were further evaluated for their promising antibacterial and anti-urease activities. In this case, the antibacterial activities were estimated by the agar well diffusion method, whereas the anti-urease activities of these compounds were determined using the indophenol method by quantifying the evolved ammonia produced. The results revealed that all the sulfacetamide derivatives displayed antibacterial activity against Bacillus subtiles, Escherichia coli, Staphylococcus aureus, Shigella dysenteriae, Salmonella typhae, Pseudomonas aeruginosa at various concentrations. Furthermore, the compound 4g N-((5-(4-chlorophenyl)thiophen-2-yl)sulfonyl) acetamide showed excellent urease inhibition with percentage inhibition activity ~46.23 ± 0.11 at 15 µg/mL with IC50 17.1 µg/mL. Moreover, some other compounds 4a-f also exhibited very good inhibition against urease enzyme.
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS