Merck
  • IFN Regulatory Factor-1 Modulates the Function of Dendritic Cells in Patients with Acute Coronary Syndrome.
IFN Regulatory Factor-1 Modulates the Function of Dendritic Cells in Patients with Acute Coronary Syndrome.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2015-05-23)
Min Guo, Rui Yan, Caixia Wang, Hongtao Shi, Meng Sun, Shuang Guo, Chuanshi Xiao
ABSTRACT
Atherosclerosis is widely recognized as a complex inflammatory disease involving pathogenic immune response of T cells and antigen-presenting cells (APCs) such as dendritic cells (DCs) and macrophages. Accumulating evidence has revealed that mature DCs play critical roles in the differentiation of effector T cells into CD4+ T cells, which effectively participate in the onset of acute coronary syndrome (ACS). IFN regulatory factor (IRF)-1 has been shown to be involved in various immune processes. The role of IRF-1 in DCs in the pathogenesis of ACS has not been investigated. We examined the relative mRNA and protein expression of IRF-1 in human monocyte-derived DCs in patients with coronary artery disease (CAD). The overexpression or silencing of IRF-1 expression in DCs in patients with ACS was performed to explore the possible role of IRF-1 in the maturation and function of DCs involved in ACS. The results showed that the relative expression of IRF-1 in DCs is obviously increased in patients with ACS. The overexpression or silencing of IRF-1 expression could effectively promote or attenuate the maturation and function of DCs. In addition, we revealed that the MAPK pathway (phosphorylation of JNK, p38 and ERK1/2) might be downstream of IRF-1 signalling pathway in activation of circulating DCs in ACS patients. The present data demonstrate that IRF-1 could effectively promote the immune maturation and function of DCs in ACS patients.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
MISSION® esiRNA, targeting human IRF1
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Interleukin-4 human, IL-4, potency:, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Thrombopoietin from mouse, recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture, >97% (SDS-PAGE)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
IL-4 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
IL-4 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Irf1
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Thrombopoietin human, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, ≥98% (SDS-PAGE and HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Interleukin-4 human, IL-4, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
SAFC
L-Glutamine