Merck
  • Nur77 Was Essential for Neurite Outgrowth and Involved in Schwann Cell Differentiation After Sciatic Nerve Injury.
Nur77 Was Essential for Neurite Outgrowth and Involved in Schwann Cell Differentiation After Sciatic Nerve Injury.
Journal of molecular neuroscience : MN (2015-05-11)
Weidong Zhang, Xudong Zhu, Yang Liu, Minhao Chen, Shixian Yan, Xingxing Mao, Zhongbing Liu, Weijie Wu, Chen Chen, Xinbao Xu, Youhua Wang
ABSTRACT
Nur77, together with Nurr1 and NOR-1, constitutes the NR4A subgroup of orphan nuclear receptors and plays critical roles in cell proliferation, differentiation, migration, and apoptosis. Among them, Nur77 is universally well known to contribute to neurite outgrowth. However, information regarding its regulation and possible function in the peripheral nervous system is still limited. In this study, we performed a sciatic nerve injury model in adult rats and detected an increased expression of Nur77 in the sciatic nerve, which was similar to the expression of Oct-6. Immunofluorescence indicated that Nur77 was located in both axons and Schwann cells. In vitro, we observed enhanced expression of Nur77 during the process of both basic fibroblast growth factor (bFGF)-induced Schwann cells differentiation and nerve growth factor (NGF)-induced PC12 cell neurite outgrowth. In vitro and in vivo experiments indicated that inhibiting the function of Nur77 by specific short hairpin RNA could depress Schwann cells myelinization and axons regeneration. Collectively, all these results suggested that upregulation of Nur77 might be involved in Schwann cells differentiation and neurite elongation following sciatic nerve crush.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Nr4a1
Sigma-Aldrich
MISSION® esiRNA, targeting human NR4A1