Merck
Graphene Quantum Dots Doping of MoS2 Monolayers.
Advanced materials (Deerfield Beach, Fla.) (2015-08-11)
Ziwei Li, Ruquan Ye, Rui Feng, Yimin Kang, Xing Zhu, James M Tour, Zheyu Fang
ABSTRACT
Graphene quantum dots (GQDs) interacting with molybdenum disulfide (MoS2 ) monolayers induce an effective photoexcited charge transfer at the interface. Both the photoluminescence (PL) and valley polarization of this GQDs/MoS2 heterostructure can be modulated under various doping charge densities. The photon-exciton interaction is used to explain and calculate the heterostructure PL control, and is further applied to the valley-polarization tuning.
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, >40 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), beads
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 35 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), ≤12 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 200 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, ≥350 μm particle size