Merck
  • High yields of fatty acid and neutral lipid production from cassava bagasse hydrolysate (CBH) by heterotrophic Chlorella protothecoides.
High yields of fatty acid and neutral lipid production from cassava bagasse hydrolysate (CBH) by heterotrophic Chlorella protothecoides.
Bioresource technology (2015-05-24)
Junhui Chen, Xiaoguang Liu, Dong Wei, Gu Chen
ABSTRACT
The fermentation process for high yields of fatty acid and neutral lipid production from cassava bagasse hydrolysate (CBH) was developed by heterotrophic Chlorella protothecoides. An efficient single-step enzymatic hydrolysis of cassava bagasse (CB) by cellulase was firstly developed to produce >30 g/L of reducing sugars. The concentrated CBH was subsequently applied in a batch culture, producing 7.9 g/L of dry biomass with yield of 0.44 g/g reducing sugar and 34.3 wt% of fatty acids and 48.6 wt% of neutral lipids. Furthermore, fed-batch fermentation using CBH achieved higher yields of fatty acids (41.0 wt% and a titer of 5.83 g/L) and neutral lipids (58.4 wt% and yield of 0.22 g/g reducing sugar). Additionally, the fatty acid profile analysis showed that the intercellular lipid was suitable to prepare high-quality biodiesel. This study demonstrated the feasibility of using CBH as low-cost feedstock to produce crude algal oil for sustainable biodiesel production.
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%