Merck
  • PAHs in the Ría de Arousa (NW Spain): A consideration of PAHs sources and abundance.
PAHs in the Ría de Arousa (NW Spain): A consideration of PAHs sources and abundance.
Marine pollution bulletin (2015-05-12)
Begoña Pérez-Fernández, Lucía Viñas, M Ángeles Franco, Jesica Bargiela
ABSTRACT
Analysis of 35 parental and alkylated homologues of PAHs (Polycyclic Aromatic Hydrocarbons) was carried out in eleven marine sediment samples collected along a central transect in the biggest Galician ría. The samples were collected using a box-core dredge and, after freeze-drying, were kept frozen until analysis. The sediments were extracted by PLE (Pressurized Liquid Extraction) procedure and the quantification of PAHs was performed using gas chromatography coupled to mass spectrometry (GC-MS) with the aid of deuterated PAH internal standards. The total concentration of studied PAHs (Σ35PAHs) ranged from 44.8 to 7901ngg(-1) dry weight (d.w.). The highest PAH concentrations were found in sediments collected near the harbour (7901ngg(-1)) and the cleanest positions were located in the outer zone of the ría. To date, these results are the first data presented in the area so they could be used for regular monitoring and control of future pollution episodes.
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Perylene, sublimed grade, ≥99.5%
Sigma-Aldrich
Perylene, ≥99%
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)