Merck
  • Ionic liquid-mediated technology to produce cellulose nanocrystals directly from wood.
Ionic liquid-mediated technology to produce cellulose nanocrystals directly from wood.
Carbohydrate polymers (2015-10-03)
Hatem Abushammala, Ingo Krossing, Marie-Pierre Laborie
ABSTRACT
We report for the first time the direct extraction of cellulose nanocrystals (CNCs) from wood by means of a 1-ethyl-3-methylimidazolium acetate ([EMIM][OAc]) treatment. A native cellulosic product could be recovered in 44% yield with respect to wood cellulose content. The product was analyzed for morphological (TEM, AFM, XRD), chemical (FTIR, (13)C CP/MAS NMR), thermal (DSC, TGA) and surface properties (Zeta potential, contact angle). These analyses evidenced the presence of partially acetylated (surface DS=0.28) nanocrystals of native cellulose I microstructure, with a crystallinity index of about 75% and aspect ratio of 65. Direct production of CNCs from wood is ascribed to the simultaneous capability of [EMIM][OAc] to (1) dissolve lignin in situ while only swelling cellulose, (2) decrease intermolecular cohesion in wood via acetylation, and (3) to catalyze cellulose hydrolysis.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Acetyl bromide, 99%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.995% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%