Merck
  • Cell cycle arrest and apoptosis induced by aspidin PB through the p53/p21 and mitochondria-dependent pathways in human osteosarcoma cells.
Cell cycle arrest and apoptosis induced by aspidin PB through the p53/p21 and mitochondria-dependent pathways in human osteosarcoma cells.
Anti-cancer drugs (2015-07-17)
Daqian Wan, Chaoyin Jiang, Xin Hua, Ting Wang, Yimin Chai
ABSTRACT
Aspidin PB is a natural product extracted from Dryopteris fragrans (L.) Schott, which has been characterized for its various biological activities. We reported that aspidin PB induced cell cycle arrest and apoptosis through the p53/p21 and mitochondria-dependent pathways in human osteosarcoma cells. Aspidin PB inhibited the proliferation of Saos-2, U2OS, and HOS cells in a dose-dependent and time-dependent manner. Aspidin PB induced changes in the cell cycle regulators (cyclin A, pRb, CDK2, p53, and p21), which caused cell cycle arrest in the S phase. We also explored the role of siRNA targeted to p53; it led to a dose-dependent attenuation of aspidin PB-induced apoptosis signaling. Moreover, after treatment with aspidin PB, the p21-silenced cells decreased significantly at the S phase. Aspidin PB increased the percentage of cells with mitochondrial membrane potential disruption. Western blot analysis showed that aspidin PB inhibited Bcl-2 expression and induced Bax expression to disintegrate the outer mitochondrial membrane and caused cytochrome C release. Mitochondrial cytochrome C release was associated with the activation of caspase-9 and caspase-3 cascades. Furthermore, the double-stranded DNA breaks and reactive oxygen species signaling were both involved in aspidin PB-induced DNA damage. In addition, aspidin PB inhibited tumor growth significantly in U2OS xenografts. Above all, we conclude that aspidin PB represents a valuable natural source and may potentially be applicable in osteosarcoma therapy.
Sigma-Aldrich
MISSION® esiRNA, targeting human KRAS
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
MISSION® esiRNA, targeting human TPT1
Sigma-Aldrich
Cyclophosphamide monohydrate, bulk package
Sigma-Aldrich
Cyclophosphamide monohydrate, ISOPAC®
Sigma-Aldrich
Rhodamine 123, mitochondrial specific fluorescent dye
Sigma-Aldrich
Rhodamine 123, BioReagent, for fluorescence, ≥85% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Nsg1
Sigma-Aldrich
MISSION® esiRNA, targeting human TP53