Merck
  • Insights into the growth of bismuth nanoparticles on 2D structured BiOCl photocatalysts: an in situ TEM investigation.
Insights into the growth of bismuth nanoparticles on 2D structured BiOCl photocatalysts: an in situ TEM investigation.
Dalton transactions (Cambridge, England : 2003) (2015-08-19)
Xiaofeng Chang, Shuangbao Wang, Qi Qi, Mohammed A Gondal, Siddique G Rashid, Si Gao, Deyuan Yang, Kai Shen, Qingyu Xu, Peng Wang
ABSTRACT
The synthetic techniques for novel photocatalytic crystals had evolved by a trial-and-error process that spanned more than two decades, and an insight into the photocatalytic crystal growth process is a challenging area and prerequisite for achieving an excellent photoactivity. Bismuth nanoparticle based hybrids, such as Bi/BiOCl composites, have recently been investigated as highly efficient photocatalytic systems because of the localized surface plasmon resonance (LSPR) of nanostructured bismuth. In this work, the observation towards the formation and growth of bismuth nanoparticles onto 2D structured BiOCl photocatalysts has been performed using a transmission electron microscope (TEM) directly in real time. The growth of bismuth nanoparticles on BiOCl nanosheets can be emulated and speeded up driven by the electron beam (e(-) beam) in TEM. The crystallinity, growth and the elemental evolution during the formation of bismuth nanoparticles have also been probed in this work.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Potassium iodide, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Potassium iodide, suitable for plant cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Potassium iodide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium iodide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Bismuth, granular, ≥99.99% trace metals basis
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Bismuth, powder, −100 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Bismuth, shot, 4-30 mesh, 99.9% trace metals basis
Sigma-Aldrich
Bismuth, beads, 1-5 mm, 99.999% trace metals basis
Sigma-Aldrich
Bismuth, pieces, 1-12 mm, 99.999% trace metals basis
Sigma-Aldrich
Bismuth, powder, −100 mesh, 99% trace metals basis
Bismuth, foil, not light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 99.97%
Bismuth, foil, not light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 99.97%
Bismuth, foil, not light tested, 100x100mm, thickness 0.025mm, permanent polyester support, 99.97%
Bismuth, foil, not light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 99.97%
Bismuth, foil, light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 99.97%
Bismuth, foil, not light tested, 50x50mm, thickness 0.001mm, permanent polyester support, 99.97%
Bismuth, foil, light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 99.97%
Bismuth, foil, 50x50mm, thickness 1.0mm, 99.97%
Bismuth, foil, light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 99.97%
Bismuth, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 99.97%
Bismuth, foil, 50x50mm, thickness 2.0mm, 100%
Bismuth, foil, 25x25mm, thickness 0.25mm, 100%