Merck
  • A versatile snap chip for high-density sub-nanoliter chip-to-chip reagent transfer.
A versatile snap chip for high-density sub-nanoliter chip-to-chip reagent transfer.
Scientific reports (2015-07-08)
Huiyan Li, Jeffrey D Munzar, Andy Ng, David Juncker
ABSTRACT
The coordinated delivery of minute amounts of different reagents is important for microfluidics and microarrays, but is dependent on advanced equipment such as microarrayers. Previously, we developed the snap chip for the direct transfer of reagents, thus realizing fluidic operations by only manipulating microscope slides. However, owing to the misalignment between arrays spotted on different slides, millimeter spacing was needed between spots and the array density was limited. In this work, we have developed a novel double transfer method and have transferred 625 spots cm(-2), corresponding to >10000 spots for a standard microscope slide. A user-friendly snapping system was manufactured to make liquid handling straightforward. Misalignment, which for direct transfer ranged from 150-250 μm, was reduced to <40 μm for double transfer. The snap chip was used to quantify 50 proteins in 16 samples simultaneously, yielding limits of detection in the pg/mL range for 35 proteins. The versatility of the snap chip is illustrated with a 4-plex homogenous enzyme inhibition assay analyzing 128 conditions with precise timing. The versatility and high density of the snap chip with double transfer allows for the development of high throughput reagent transfer protocols compatible with a variety of applications.
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Trichloro(1H,1H,2H,2H-perfluorooctyl)silane, 97%
Sigma-Aldrich
(±)-1,3-Butanediol, anhydrous, ≥99%
Sigma-Aldrich
Leupeptin hydrochloride, microbial, ≥70% (HPLC)
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent
Sigma-Aldrich
Leupeptin hydrochloride, microbial, ≥90% (HPLC)
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
(±)-1,3-Butanediol, ReagentPlus®, 99.5%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Betaine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)