Merck
  • Impact of Meyerozyma guilliermondii isolated from chickens against Eimeria sp. protozoan, an in vitro analysis.
Impact of Meyerozyma guilliermondii isolated from chickens against Eimeria sp. protozoan, an in vitro analysis.
BMC veterinary research (2015-11-11)
Edgar Dantán-González, Rosa Estela Quiroz-Castañeda, Mayra Cobaxin-Cárdenas, Jorge Valle-Hernández, Yitzel Gama-Martínez, José Raunel Tinoco-Valencia, Leobardo Serrano-Carreón, Laura Ortiz-Hernández
ABSTRACT
Avian coccidiosis is a disease caused worldwide by several species of parasite Eimeria that causes significant economic losses. This disease affects chickens development and production, that most of times is controlled with anticoccidial drugs. Although efforts have been made to address this disease, they have been made to control Eimeria sporozoites, although enteric stages are often vulnerable, however; the parasite oocyst remains a problem that must be controlled, as it has a resistant structure that facilitates dispersion. Despite some commercial products based on chemical compounds have been developed as disinfectants that destroy oocysts, the solution of the problem remains to be solved. In this work, we assessed in vitro anticoccidial activity of a compound(s) secreted by yeast isolated in oocysts suspension from infected chickens. The yeast was molecularly identified as Meyerozyma guilliermondii, and its anticoccidial activity against Eimeria tenella oocysts was assessed. Here, we report the damage to oocysts walls caused by M. guilliermondii culture, supernatant, supernatant extract and intracellular proteins. In all cases, a significant decreased of oocysts was observed. The yeast Meyerozyma guilliermondii secretes a compound with anticoccidial activity and also has a compound of protein nature that damages the resistant structure of oocyst, showing the potential of this yeast and its products as a feasible method of coccidiosis control.
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis