Merck
  • In vitro permeability across Caco-2 cells (colonic) can predict in vivo (small intestinal) absorption in man--fact or myth.
In vitro permeability across Caco-2 cells (colonic) can predict in vivo (small intestinal) absorption in man--fact or myth.
Pharmaceutical research (1997-06-01)
S Yee
ABSTRACT
To evaluate and optimize the use of Caco-2 cell monolayers to predict the in vivo absorption of a broad range of compounds in man. Caco-2 cells are derived from human adenocarcinoma colon cells and spontaneously differentiate when grown on porous polyethylene terephthalate membranes (PETP) in a 12 well format to form monolayers of polarized cells possessing function similar to intestinal enterocytes. Transport experiments were conducted using 21 day cultured cells in a shaking water bath at 37 degrees C. Radiolabeled mannitol was used to determine monolayer integrity. Apparent permeability coefficient (Papp) was calculated from the appearance of drug in the receiver side. A strong correlation was observed between in vivo human absorption and in vitro Papp for a variety of compounds (R = 0.95, N = 35). For compounds that are substrates of p-glycoprotein (Pgp), use of a Pgp inhibitor resulted in a better estimate of absorption in humans. The results of this study suggest that the overall ranking of compounds with Papp < 1 x 10(-6) cm/sec, between 1-10 x 10(-5) cm/ sec, and > 10 x 10(-6) cm/sec can be classified as poorly (0-20%), moderately (20-70%) and well (70-100%) absorbed compounds, respectively. These data suggest that Caco-2 cells developed under the culturing and transport conditions defined herein can be used to predict in vivo human absorption of compounds regardless of transport mechanism, viz., transcellular, paracellular and carrier-mediated.
Sigma-Aldrich
MDR1 Knockout Caco-2 Cells, one vial
Sigma-Aldrich
MDR1/MRP2 Double Knockout Caco-2 Cells, one vial
Sigma-Aldrich
MRP4 Knockout Caco-2 Cells, one vial
Sigma-Aldrich
MRP5 Knockout Caco-2 Cells, one vial
Sigma-Aldrich
MRP7 Knockout Caco-2 Cells, one vial
Sigma-Aldrich
MRP1 Knockout Caco-2 Cells, one vial
Sigma-Aldrich
Caco-2 Engineered Control Cells, one vial of frozen cells
Sigma-Aldrich
MDR1/BCRP Double Knockout Caco-2 Cells, one vial
Sigma-Aldrich
MRP2 Knockout Caco-2 Cells, one vial
Sigma-Aldrich
BCRP/MRP2 Double Knockout Caco-2 Cells, one vial
Sigma-Aldrich
BCRP Knockout Caco-2 Cells, one vial
Sigma-Aldrich
MRP3 Knockout Caco-2 Cells, one vial