Merck
  • A single dose of EGLN1 siRNA yields increased erythropoiesis in nonhuman primates.
A single dose of EGLN1 siRNA yields increased erythropoiesis in nonhuman primates.
Nucleic acid therapeutics (2014-10-02)
Marc T Abrams, Martin Koser, Julja Burchard, Walter Strapps, Huseyin Mehmet, Marian Gindy, Dennis Zaller, Laura Sepp-Lorenzino, Dominique Stickens
ABSTRACT
Decreased production of erythropoietin (EPO) causes anemia in patients with chronic kidney disease, and recombinant human EPO is used to treat renal failure associated anemia. The liver, the main EPO-producing organ in utero, maintains the capacity to produce EPO in the adult but in insufficient quantities to restore hemoglobin levels to normal in patients with impaired renal function. Inhibition of prolyl-4-hydroxylase domain (PHD) proteins is known to cause an increase in EPO production through its effects on hypoxia inducible factor. Here, we utilized small interfering RNA (siRNA) targeting EGLN1, the gene encoding the PHD2 protein, to investigate the phenotypic consequences in nonhuman primates. A single, well-tolerated intravenous dose of an optimized EGLN1 siRNA encapsulated in a lipid nanoparticle formulation caused robust mRNA silencing in the liver, leading to increases in serum EPO and hemoglobin. The siRNA-induced erythropoiesis was dose-dependent and was sustained for at least 2 months. These data point to the potential for an RNA interference-based, liver-targeted therapeutic approach for the treatment of anemia.
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethanol, for residue analysis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications