Merck
  • Investigation of antibody disulfide reduction and re-oxidation and impact to biological activities.
Investigation of antibody disulfide reduction and re-oxidation and impact to biological activities.
Journal of pharmaceutical and biomedical analysis (2014-12-03)
Tian Wang, Yaoqing Diana Liu, Bing Cai, Gang Huang, Gregory C Flynn
ABSTRACT
Disulfide reduction in therapeutic monoclonal antibodies can occur during cell harvest operations as a result of cell breakage. Understanding these product quality changes and manufacturers' ability to control them would likely be of concern to regulatory bodies. To study the biological impact of disulfide reduction, mAbs, including IgG2κ, IgG2λ, IgG1κ, and IgG1λ forms, were partially reduced with dithiothreitol (DTT). Samples generated had approximately 10% or 50% intact molecules as determined by nrCE-SDS. Similar to the type of partial reduction obtained during uncontrolled harvest operations, DTT reduced antibodies were free from sulfur-linked adduct, such as attached cysteine. These partially reduced materials were incubated under physiological (blood-mimicking) redox conditions in vitro to follow the fate of the interchain cysteines. Within 8h, the original disulfide bonds reformed. For mAbA, an IgG2κ, the initial re-oxidized state favored the IgG2-A disulfide isoform, which then underwent conversion over time to other isoforms. Reduced material was fully active. Results suggest that the type of disulfide reduction would have minimal impact to safety or efficacy. Antibody re-oxidation rates were found to be in the order of IgG2κ<IgG2λ<IgG1κ and IgG1λ, the same order as previously determined reduction rates.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
SAFC
L-Cystine
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
L-Cystine, ≥99.7% (TLC)
Sigma-Aldrich
2-Nitrobenzoic acid, 95%, Contains 3- and 4-isomers
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
Guanidine hydrochloride solution, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
L-Cystine, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
L-Cystine, ≥98% (TLC), crystalline
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
L-Cystine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
L-Cystine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.5%