Merck
  • The small GTPase Rap1 promotes cell movement rather than stabilizes adhesion in epithelial cells responding to insulin-like growth factor I.
The small GTPase Rap1 promotes cell movement rather than stabilizes adhesion in epithelial cells responding to insulin-like growth factor I.
The Biochemical journal (2014-07-17)
Marina A Guvakova, William S Y Lee, Dana K Furstenau, Indira Prabakaran, David C Li, Rupert Hung, Natasha Kushnir
ABSTRACT
The Ras-related GTPase Rap1 promotes cell adhesion and migration. Although the significance of Rap1 contribution to cell migration is increasingly being recognized, little is known about the biochemical mechanisms driving this process. In the present study, we discovered a previously unidentified regulatory role of insulin-like growth factor type I (IGF-I) receptor (IGF-IR) in CRK Src homology 3 (SH3)-binding guanine-nucleotide-releasing protein (C3G)-Rap1-fascin-actin axis promoting cell movement. We demonstrate that a burst of Rap1 activity, rather than presumed hyperactivation, is imperative for the onset of cell movement. We show that while autophosphorylated IGF-IR signals to C3G to activate Rap1, subsequent IGF-IR internalization promotes gradual inactivation of Rap1 by putative Rap1 GTPase-activating protein (GAP). Additionally, IGF-IR signalling recruits active Rap1 at sites of cell motile protrusions. C3G depletion prevents IGF-I-induced fascin accumulation at actin microspikes and blocks protrusions. In the absence of IGF-IR activity, the wild-type (WT) Rap1 and the constitutively active V12Rap1 mutant remain in cell-cell contacts. Forced inactivation of Rap1 signalling by overexpressing dominant negative N17Rap1, Rap1GAP or by silencing C3G has a detrimental effect on filamentous (F)-actin and cell adhesion irrespective of IGF-IR signalling. We conclude that the basal levels of Rap1 activity holds up cell adhesion, whereas sequential regulation of C3G and GAP by IGF-IR reverses the labile Rap1 function from supporting adhesion to promoting migration.
Sigma-Aldrich
MISSION® esiRNA, targeting human RAPGEF1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Rapgef1
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Thrombopoietin human, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, ≥98% (SDS-PAGE and HPLC)
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
tert-Butyl acetoacetate, reagent grade, 98%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dansylcadaverine, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
Dansylcadaverine, ≥97% (TLC)
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard