Merck
  • Four basic residues critical for the ion selectivity and pore blocker sensitivity of TMEM16A calcium-activated chloride channels.
Four basic residues critical for the ion selectivity and pore blocker sensitivity of TMEM16A calcium-activated chloride channels.
Proceedings of the National Academy of Sciences of the United States of America (2015-03-04)
Christian J Peters, Haibo Yu, Jason Tien, Yuh Nung Jan, Min Li, Lily Yeh Jan
ABSTRACT
TMEM16A (transmembrane protein 16) (Anoctamin-1) forms a calcium-activated chloride channel (CaCC) that regulates a broad array of physiological properties in response to changes in intracellular calcium concentration. Although known to conduct anions according to the Eisenman type I selectivity sequence, the structural determinants of TMEM16A anion selectivity are not well-understood. Reasoning that the positive charges on basic residues are likely contributors to anion selectivity, we performed whole-cell recordings of mutants with alanine substitution for basic residues within the putative pore region and identified four residues on four different putative transmembrane segments that significantly increased the permeability of the larger halides and thiocyanate relative to that of chloride. Because TMEM16A permeation properties are known to shift with changes in intracellular calcium concentration, we further examined the calcium dependence of anion selectivity. We found that WT TMEM16A but not mutants with alanine substitution at those four basic residues exhibited a clear decline in the preference for larger anions as intracellular calcium was increased. Having implicated these residues as contributing to the TMEM16A pore, we scrutinized candidate small molecules from a high-throughput CaCC inhibitor screen to identify two compounds that act as pore blockers. Mutations of those four putative pore-lining basic residues significantly altered the IC50 of these compounds at positive voltages. These findings contribute to our understanding regarding anion permeation of TMEM16A CaCC and provide valuable pharmacological tools to probe the channel pore.
Sigma-Aldrich
1PBC, ≥98% (HPLC)
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Calcium, granular, 99%
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Calcium, turnings, 99% trace metals basis
Sigma-Aldrich
Calcium, pieces, <1 cm, 99%