MilliporeSigma
  • The histone acetylranseferase hMOF acetylates Nrf2 and regulates anti-drug responses in human non-small cell lung cancer.
The histone acetylranseferase hMOF acetylates Nrf2 and regulates anti-drug responses in human non-small cell lung cancer.
British journal of pharmacology (2014-02-28)
Zhiwei Chen, Xiangyun Ye, Naiwang Tang, Shengping Shen, Ziming Li, Xiaomin Niu, Shun Lu, Ling Xu
ABSTRACT
The histone acetyltransferase MOF is a member of the MYST family. In mammals, MOF plays critical roles by acetylating histone H4 at K16 and non-histone substrates such as p53. Here we have investigated the role of MOF in human lung cancer and possible new substrates of hMOF. Samples of human non-small cell lung cancer (NSCLC) were used to correlate MOF with clinicopathological parameters and NF-E2-related factor 2 (Nrf2) downstream genes. 293T-cells were used to study interactions between MOF and Nrf2, and acetylation of Nrf2 by MOF. Mouse embryonic fibroblast and A549 cells were utilized to assess involvement of MOF in antioxidative and anti-drug responses. A549 cells were used to analysis the role of MOF in anti-drug response in vitro and in vivo. hMOF was overexpressed in human NSCLC tissues and was associated with large tumour size, advanced disease stage and metastasis, and with poor prognosis. hMOF levels were positively correlated with Nrf2-downstream genes. MOF/hMOF physically interacted with and acetylated Nrf2 at Lys(588) . MOF-mediated acetylation increased nuclear retention of Nrf2 and transcription of its downstream genes. Importantly, MOF/hMOF was essential for anti-oxidative and anti-drug responses in vitro and regulated tumour growth and drug resistance in vivo in an Nrf2-dependent manner. hMOF was overexpressed in human NSCLC and was a predictor of poor survival. hMOF-mediated Nrf2 acetylation and nuclear retention are essential for anti-oxidative and anti-drug responses. hMOF may provide a therapeutic target for the treatment of NSCLC.
Product Description
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
SKU
Pack Size
Availability
Price
Quantity
Fluorouracil, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
SKU
Pack Size
Availability
Price
Quantity
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
SKU
Pack Size
Availability
Price
Quantity
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
SKU
Pack Size
Availability
Price
Quantity
Supelco
5-Fluorouracil, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
2′,7′-Dichlorodihydrofluorescein diacetate, ≥97%
SKU
Pack Size
Availability
Price
Quantity
USP
Fluorouracil, United States Pharmacopeia (USP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MISSION® esiRNA, targeting human KAT8
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Myst1
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
SKU
Pack Size
Availability
Price
Quantity
Cisplatin, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline