Merck
  • New co-processed MCC-based excipient for fast release of low solubility drugs from pellets prepared by extrusion-spheronization.
New co-processed MCC-based excipient for fast release of low solubility drugs from pellets prepared by extrusion-spheronization.
Drug development and industrial pharmacy (2013-11-28)
Alvaro Goyanes, Ramón Martínez-Pacheco
ABSTRACT
In this study, a new co-processed excipient composed of microcrystalline cellulose (MCC), sorbitol, chitosan and Eudragit® E, easily obtained by wet massing, to increase the dissolution rate of active ingredients of low water solubility from pellets prepared by extrusion-spheronization is evaluated. Indomethacin, nifedipine, furosemide, ibuprofen, prednisolone and hydrochlorothiazide are used as model drugs of different solubility. All pellet formulations evaluated showed adequate morphological, flow and mechanical properties. Pellets prepared with the co-processed excipient show a higher drug dissolution rate than those prepared with MCC and even higher than the pure drug powder. The fast drug dissolution and the complete disintegration (<3 min) of the pellets can be explained by the great porosity of the formulations, the high solubility of the sorbitol, the disintegrant capacity of the chitosan and the distribution of the Eudragit® E polymer particles in-between the other components of the co-processed mixture. In conclusion, this new co-processed excipient is very suitable to increase the dissolution rate of poorly soluble drugs from pellets prepared by extrusion-spheronization. Moreover, the drug release rate can be estimated from the Ln of the drug solubility in acidic medium.
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), for molecular biology
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Sorbitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sorbitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
USP
Sorbitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%