Merck
  • Plantago lanceolata L. water extract induces transition of fibroblasts into myofibroblasts and increases tensile strength of healing skin wounds.
Plantago lanceolata L. water extract induces transition of fibroblasts into myofibroblasts and increases tensile strength of healing skin wounds.
The Journal of pharmacy and pharmacology (2014-09-23)
Ivan Kováč, Ján Ďurkáč, Martin Hollý, Katarína Jakubčová, Vlasta Peržeľová, Pavel Mučaji, Emil Švajdlenka, František Sabol, Jaroslav Legáth, Jozef Belák, Karel Smetana, Peter Gál
ABSTRACT
Although the exact underlying mechanisms are still unknown, Plantago lanceolata L. (PL) water extracts are frequently used to stimulate wound healing and to drain abscesses. Therefore, in this experimental study the effect of PL water extract on skin wound healing was studied in Sprague-Dawley rats. Two excisional and one incisional skin wounds were performed on the back of each rat. Wounds were treated for three consecutive days with two different concentrations of the aqueous extract of PL. Rats were sacrificed 7, 14, and 21 days after surgery. Samples of wounds were processed for macroscopic (excisions - wound contraction measurement), biomechanical (incisions - wound tensile strength (TS) measurement) and histological examination (excisions). It was shown that open wounds treated with PL extract contained myofibroblasts and demonstrated significantly higher contraction rates. Furthermore, significantly increased wound TSs were recorded in treated rats as a consequence of increased organization of extracellular matrix proteins, such as the collagen type 1. We demonstrated that PL aqueous extract improves skin wound healing in rats. However, further research need to be performed to find optimal therapeutic concentration, and exact underlying mechanism prior obtained results may be introduced into the clinical practice.
Product Description
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Echinacoside, 98% (HPLC)
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Supelco
L-Lysine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
DAPI, for nucleic acid staining
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Echinacoside, analytical standard
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%