Merck
  • Preparation of concentrated stable fenofibrate suspensions via liquid antisolvent precipitation.
Preparation of concentrated stable fenofibrate suspensions via liquid antisolvent precipitation.
Drug development and industrial pharmacy (2013-10-10)
Mohammad A Azad, Catharina Knieke, Daniel To, Rajesh Davé
ABSTRACT
A major challenge in achieving size stability for relatively high concentration of fine particles from poorly water-soluble drug fenofibrate (FNB) is addressed through T-mixing based liquid antisolvent precipitation in the presence of ultrasonication and judicious use of stabilizers. Multiple stabilizers were screened in a batch mode prior to their continuous formation via T-mixing. In both cases, the stable suspensions maintained their size after 2 days of storage at room temperature, with the smallest particle size of d50: ∼1.2 µm was achieved through a combination of HPMC with SDS or PF-68. The influence of processing parameters, such as sonication energy, sonication probe insert depth and solvent/antisolvent flow rate, on the particle size distribution (PSD) in T-mixing were investigated, to identify optimum processing conditions. Optimal operating and formulation conditions also allowed increase in the drug loading from 0.32% to 4% (w/v), while keeping the median size 2.5 µm. Interestingly, the primary particles observed in the SEM were spherical and under 100 nm in diameter, indicating agglomeration. It was shown that the stabilized particles could be centrifuged and did not show size growth upon resuspension, allowing for increase in the drug loading up to 27% (w/v), which is a significant novel outcome.
Product Description
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Docusate sodium salt, BioUltra, ≥99.0% (TLC)
Sigma-Aldrich
Dioctyl sulfosuccinate sodium salt, ≥97%
Sigma-Aldrich
Docusate sodium, meets USP testing specifications
Sigma-Aldrich
Docusate sodium salt, BioXtra, ≥99%
Sigma-Aldrich
Docusate sodium salt, purum, ≥96.0% (TLC)