Merck
  • Molecular markers in ambient aerosol in the Mahanadi Riverside Basin of eastern central India during winter.
Molecular markers in ambient aerosol in the Mahanadi Riverside Basin of eastern central India during winter.
Environmental science and pollution research international (2014-08-19)
Jayant Nirmalkar, Manas K Deb, Dhananjay K Deshmukh, Ying I Tsai, Santosh K Verma
ABSTRACT
Organic molecular markers are important atmospheric constituents. Their formation and sources are important aspects of the study of urban and rural air quality. We collected PM10 aerosol samples from the Mahanadi Riverside Basin (MRB), a rural part of eastern central India, during the winter of 2011. PM10 aerosols were characterized for molecular markers using ion chromatography. The concentration of PM10 ranged from 208.8 to 588.3 μg m(-3) with a mean concentration of 388.9 μg m(-3). Total concentration of anhydrosugars, sugar alcohols, primary sugars, and oxalate were found to be 3.25, 5.60, 10.52, and 0.37 μg m(-3), respectively, during the study period. Glucose was the most abundant species followed by levoglucosan and mannitol. Significant positive correlation between the molecular markers, anhydrosugars, sugar alcohols, primary sugars, and oxalic acid confirmed that biomass burning, biogenic activity, and re-suspension of soil particles were the main sources of aerosol in the eastern central India study area.
Sigma-Aldrich
Xylitol, ≥99% (GC)
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)
Sigma-Aldrich
D-(+)-Galactose, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC)
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
myo-Inositol, ≥99%
Sigma-Aldrich
myo-Inositol, ≥99% (GC), BioReagent
Millipore
D-(+)-Mannose, ≥99.0% (sum of enantiomers, HPLC), suitable for microbiology
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Millipore
myo-Inositol, ≥99.0%, suitable for microbiology
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Millipore
D-(+)-Mannose, suitable for microbiology, ≥99%
Millipore
D-(+)-Galactose, suitable for microbiology, ≥99.0%
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Xylitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
USP
Xylitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99% (GC)