Merck
  • Effects of erythromycin and rifampicin on immunomodulatory gene expression and cellular function in human polymorphonuclear leukocytes.
Effects of erythromycin and rifampicin on immunomodulatory gene expression and cellular function in human polymorphonuclear leukocytes.
Chemotherapy (2014-05-24)
Xiaoqin Mu, Tsuneyuki Ubagai, Takane Kikuchi-Ueda, Shigeru Tansho-Nagakawa, Ryuichi Nakano, Hirotoshi Kikuchi, Yasuo Ono
ABSTRACT
We investigated the effects of two antibiotics, erythromycin and rifampicin, on the immunomodulatory gene expression and cellular function of human polymorphonuclear leukocytes (PMNs). We used real-time quantitative PCR to examine the expression of immunomodulatory genes. The production of reactive oxygen species (ROS) was determined by fluorescence-activated cell sorting. PMN chemotaxis was analyzed using a KK chemotaxis chamber. Stimulation of PMNs with lipopolysaccharide (LPS) resulted in increases in the mRNA levels of immunomodulatory genes. Rifampicin significantly inhibited the overexpression of TLR2, TLR4, CD14 and IL8Rs. However, erythromycin suppressed only the upregulation of TLR2 and TNFA. Neither antibiotic had an effect on the production of ROS. Rifampicin significantly inhibited PMN chemotaxis, but erythromycin had no effect. Erythromycin and rifampicin may play anti-inflammatory roles by affecting the expression levels of immunomodulatory genes or the chemotaxis of PMNs.
Product Description
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Erythromycin, for microbiological assay, European Pharmacopoeia (EP) Reference Standard
USP
Erythromycin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Rifampicin, European Pharmacopoeia (EP) Reference Standard