Merck
  • In situ electrochemical assessment of cytotoxicity of chlorophenols in MCF-7 and HeLa cells.
In situ electrochemical assessment of cytotoxicity of chlorophenols in MCF-7 and HeLa cells.
Analytical biochemistry (2014-06-29)
Hongwei Qin, Jiguang Liu, Zeshi Zhang, Jinlian Li, Guanggang Gao, Yuxin Yang, Xing Yuan, Dongmei Wu
ABSTRACT
An in situ electrochemical method was used to assess the cytotoxicity of chlorophenols using human breast cancer (MCF-7) and cervical carcinoma (HeLa) cells as models. On treatment with different chlorophenols, the electrochemical responses of the selected cells, resulting from the oxidation of guanine and xanthine in the cytoplasm, indicated the cell viability. In addition, the in situ in vitro electrochemical method was further compared with the traditional MTT (3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide) assays. Although similar cytotoxicity data were obtained from both methods, the effective concentrations of chlorophenols that inhibited 50% cell growth (EC50 values) from the electrochemical method were only slightly lower than those from the MTT assay. These results indicate that the in situ in vitro electrochemical method paves a simple, rapid, strongly responsive, and label-free way to the cytotoxicity assessment of different chlorophenol pollutants.
Sigma-Aldrich
Pentachlorophenol, 97%
Supelco
Guanine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Guanine, 98%
Sigma-Aldrich
Guanine, BioUltra
Supelco
Pentachlorophenol, analytical standard
Sigma-Aldrich
2,4-Dichlorophenol, 99%
Sigma-Aldrich
Xanthine, BioUltra, ≥99%
Sigma-Aldrich
Xanthine, ≥99.5% (HPLC), purified by recrystallization
Sigma-Aldrich
Xanthine, ≥99%
Sigma-Aldrich
2,4,6-Trichlorophenol, 98%