Merck
  • Enhanced cellular uptake and gene silencing activity of siRNA molecules mediated by chitosan-derivative nanocomplexes.
Enhanced cellular uptake and gene silencing activity of siRNA molecules mediated by chitosan-derivative nanocomplexes.
International journal of pharmaceutics (2014-07-27)
Diana Guzman-Villanueva, Ibrahim M El-Sherbiny, Alexander V Vlassov, Dea Herrera-Ruiz, Hugh D C Smyth
ABSTRACT
The RNA interference (RNAi) constitutes a conservative mechanism in eukaryotic cells that induces silencing of target genes. In mammalians, the RNAi is triggered by siRNA (small interfering RNA) molecules. Due to its potential in silencing specific genes, the siRNA has been considered a potential alternative for the treatment of genetic and acquired diseases. However, the siRNA therapy has been limited by its low stability and rapid degradation in presence of nucleases, low cellular uptake, and immune response activation. In order to overcome these drawbacks, we propose the synthesis and characterization of non-viral delivery systems using chitosan derivatives to obtain siRNA complexes (polyplexes). The non-viral delivery systems synthesized included PEG-g-OCs (oligochitosan) and PEG-g-Cs (chitosan medium molecular weight). Both systems allowed the formation of siRNA polyplexes, increased the stability of siRNA in the presence of nucleases, enhanced cellular internalization, and showed low toxicity in the A549 cell line. Finally, the complexes obtained with the PEG-g-OCs system showed silencing activity in a GFP model in the cell line A549 in comparison with naked siRNA.
Product Description
Valaciclovir impurity G, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
1,4-Dioxane, ACS reagent, ≥99.0%
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
1,4-Dioxane, ACS reagent, ≥99.0%, contains ≤25 ppm BHT as stabilizer
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
1,4-Dioxane, suitable for HPLC, ≥99.5%
Supelco
N,N-Dimethylformamide, analytical standard
USP
Valacyclovir Related Compound G, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Hydrazine hydrate, reagent grade, N2H4 50-60 %
Sigma-Aldrich
Phthalic anhydride, ReagentPlus®, 99%
Sigma-Aldrich
Phthalic anhydride, ACS reagent, ≥99%
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Triethylamine, ≥99%
Supelco
Triethylamine, analytical standard
Supelco
1,4-Dioxane, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
1,4-Dioxane, analytical standard
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
4-(Dimethylamino)pyridine, purum, ≥98.0% (NT)
Sigma-Aldrich
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydrazine monohydrate, N2H4 64-65 %, reagent grade, ≥97%
Sigma-Aldrich
Hydrazine hydrate solution, puriss. p.a., 24-26% in H2O (RT)