Merck
  • Mutations in the chikungunya virus non-structural proteins cause resistance to favipiravir (T-705), a broad-spectrum antiviral.
Mutations in the chikungunya virus non-structural proteins cause resistance to favipiravir (T-705), a broad-spectrum antiviral.
The Journal of antimicrobial chemotherapy (2014-06-22)
Leen Delang, Nidya Segura Guerrero, Ali Tas, Gilles Quérat, Boris Pastorino, Mathy Froeyen, Kai Dallmeier, Dirk Jochmans, Piet Herdewijn, Felio Bello, Eric J Snijder, Xavier de Lamballerie, Byron Martina, Johan Neyts, Martijn J van Hemert, Pieter Leyssen
ABSTRACT
T-705, also known as favipiravir, is a small-molecule inhibitor that is currently in clinical development for the treatment of influenza virus infections. This molecule also inhibits the replication of a broad spectrum of other RNA viruses. The objective of this study was to investigate the antiviral effect of favipiravir on chikungunya virus (CHIKV) replication and to contribute to unravelling the molecular mechanism of action against this virus. The anti-CHIKV effect of favipiravir was examined in cell culture and in a mouse model of lethal infection. A five-step protocol was used to select for CHIKV variants with reduced susceptibility to favipiravir. The resistant phenotype was confirmed in cell culture and the whole genome was sequenced. The identified mutations were reverse-engineered into an infectious clone to confirm their impact on the antiviral efficacy of favipiravir. Favipiravir inhibits the replication of laboratory strains and clinical isolates of CHIKV, as well as of a panel of other alphaviruses. Several favipiravir-resistant CHIKV variants were independently selected and all of them in particular acquired the unique K291R mutation in the RNA-dependent RNA polymerase (RdRp). Reverse-engineering of this K291R mutation into an infectious clone of CHIKV confirmed the link between the mutant genotype and the resistant phenotype. Interestingly, this particular lysine is also highly conserved in the RdRp of positive-stranded RNA viruses in general. This study provides an important insight into the precise molecular mechanism by which favipiravir exerts its antiviral activity against (alpha)viruses, which may be of help in designing other potent broad-spectrum antivirals.
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Lithium chloride, powder, ≥99.98% trace metals basis
Supelco
Lithium chloride solution, 1 M in ethanol
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)
Sigma-Aldrich
Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Lithium chloride, BioUltra, for molecular biology, anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
Sigma-Aldrich
Lithium chloride, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Lithium chloride solution, 8 M, for molecular biology, ≥99%
Sigma-Aldrich
Lithium chloride, for molecular biology, ≥99%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Lithium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Lithium chloride, ReagentPlus®, 99%
Sigma-Aldrich
Lithium chloride, ACS reagent, ≥99%
Supelco
Electrolyte solution, nonaqueous, LiCl in ethanol (saturated)
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Electrolyte solution, nonaqueous, 2 M LiCl in ethanol
Sigma-Aldrich
L-Glutamine