Merck
  • The effect of AMA0428, a novel and potent ROCK inhibitor, in a model of neovascular age-related macular degeneration.
The effect of AMA0428, a novel and potent ROCK inhibitor, in a model of neovascular age-related macular degeneration.
Investigative ophthalmology & visual science (2015-01-30)
Karolien Hollanders, Tine Van Bergen, Nele Kindt, Karolien Castermans, Dirk Leysen, Evelien Vandewalle, Lieve Moons, Ingeborg Stalmans
ABSTRACT
Rho kinase (ROCK) is associated with VEGF-driven angiogenesis, as well as with inflammation and fibrosis. Therefore, the effect of AMA0428, a novel ROCK inhibitor, was studied in these processes, which highly contribute to the pathogenesis of neovascular AMD. The effect of AMA0428 (0.5-5.0 μM) on human umbilical vein endothelial cells (HUVECs), human brain microvascular endothelial cells (HBMECs), and human brain microvascular pericytes (HBVPs) was determined using cell viability (WST-1), apoptosis (caspase 3/7), and migration (scratch and under-agarose) assays. The in vivo response was investigated using a laser-induced choroidal neovascularization (CNV) mouse model, in which intravitreal injections of AMA0428, murine anti-VEGF-R2 mAb (DC101), or placebo was given. Outcome was assessed by analysis of inflammation (CD45), angiogenesis (FITC-dextran), vessel leakage (Texas Red-conjugated Dextran and FITC-labeled lectin) and fibrosis (Sirius Red/Collagen I). The AMA0428 dose-dependently reduced proliferation and VEGF-induced migration of HUVEC and HBMEC (P < 0.05). No significant effect was seen on HBVP proliferation; however, migration and pericyte recruitment were enhanced (P < 0.05) by AMA0428 administration. There was no apoptosis induction. The AMA0428 significantly reduced CNV and vessel leakage 2 weeks after laser treatment, comparable to DC101. In addition, AMA0428 inhibited inflammation on day 5 by 42% (P < 0.05) and collagen deposition on day 30 by 43% (P < 0. 05), whereas DC101 had no effect on inflammation nor on fibrosis. The results suggest that targeting ROCK with AMA0428 not only reduces neoangiogenesis, but also blocks inflammation and fibrosis (contrary to VEGF suppression). These results point to a potential therapeutic benefit of ROCK inhibition in neovascular AMD.
Product Description
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Picric acid solution, 1.3% in H2O (saturated)
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – 2-propanol solution, derivatization grade (GC derivatization), ~1.25 M HCl (T), LiChropur
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Vascular Endothelial Growth Factor human, VEGF, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture