Merck
  • Protein corona of nanoparticles: distinct proteins regulate the cellular uptake.
Protein corona of nanoparticles: distinct proteins regulate the cellular uptake.
Biomacromolecules (2015-03-21)
Sandra Ritz, Susanne Schöttler, Niklas Kotman, Grit Baier, Anna Musyanovych, Jörg Kuharev, Katharina Landfester, Hansjörg Schild, Olaf Jahn, Stefan Tenzer, Volker Mailänder
ABSTRACT
Understanding nanoparticle-protein interactions is a crucial issue in the development of targeted nanomaterial delivery. Besides unraveling the composition of the nanoparticle's protein coronas, distinct proteins thereof could control nanoparticle uptake into specific cell types. Here we differentially analyzed the protein corona composition on four polymeric differently functionalized nanoparticles by label-free quantitative mass spectrometry. Next, we correlated the relative abundance of identified proteins in the corona with enhanced or decreased cellular uptake of nanoparticles into human cancer and bone marrow stem cells to identify key candidates. Finally, we verified these candidate proteins by artificially decorating nanoparticles with individual proteins showing that nanoparticles precoated with the apolipoproteins ApoA4 or ApoC3 significantly decreased the cellular uptake, whereas precoating with ApoH increased the cellular uptake.
Product Description
Supelco
Ethylene oxide solution, certified reference material, 50 mg/mL in methanol
Sigma-Aldrich
Acrylic acid, anhydrous, contains 200 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Hexadecane, anhydrous, ≥99%
Sigma-Aldrich
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethylene oxide solution, 2.5-3.3 M in THF
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Styrene, analytical standard
Supelco
Hexadecane, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Hexadecane, ReagentPlus®, 99%
Sigma-Aldrich
2-Aminoethyl methacrylate hydrochloride, contains ~500 ppm phenothiazine as stabilizer, 90%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Zirconyl chloride octahydrate, reagent grade, 98%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Ethylene oxide, ≥99.5%
Sigma-Aldrich
Sodium 4-vinylbenzenesulfonate, technical, ≥90% (T)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)