Merck
  • Convenient synthesis of 18F-radiolabeled R-(-)-N-n-propyl-2-(3-fluoropropanoxy-11-hydroxynoraporphine.
Convenient synthesis of 18F-radiolabeled R-(-)-N-n-propyl-2-(3-fluoropropanoxy-11-hydroxynoraporphine.
Journal of labelled compounds & radiopharmaceuticals (2014-11-18)
Anna W Sromek, Shaohui Zhang, Vamsidhar Akurathi, Alan B Packard, Wei Li, David Alagille, Thomas J Morley, Ronald Baldwin, Gilles Tamagnan, John L Neumeyer
ABSTRACT
Aporphines are attractive candidates for imaging D2 receptor function because, as agonists rather than antagonists, they are selective for the receptor in the high affinity state. In contrast, D2 antagonists do not distinguish between the high and low affinity states, and in vitro data suggests that this distinction may be important in studying diseases characterized by D2 dysregulation, such as schizophrenia and Parkinson's disease. Accordingly, MCL-536 (R-(-)-N-n-propyl-2-(3-[(18)F]fluoropropanoxy-11-hydroxynoraporphine) was selected for labeling with (18)F based on in vitro data obtained for the non-radioactive ((19)F) compound. Fluorine-18-labeled MCL-536 was synthesized in 70% radiochemical yield, >99% radiochemical purity, and specific activity of 167 GBq/µmol (4.5 Ci/µmol) using p-toluenesulfonyl (tosyl) both as a novel protecting group for the phenol and a leaving group for the radiofluorination.
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Potassium fluoride, ≥99.97% trace metals basis
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
tert-Butylchlorodimethylsilane solution, 50 wt. % in toluene
Supelco
Acetone, analytical standard
Sigma-Aldrich
Phosphorus pentoxide, desiccant, with moisture indicator
Supelco
Tetrahydrofuran, analytical standard
Sigma-Aldrich
Phosphorus pentoxide, puriss. p.a., ACS reagent, ≥98.0% (T)
Supelco
Methanol, analytical standard
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Phosphorus pentoxide, ReagentPlus®, 99%
Supelco
Phosphorus pentoxide, ≥99.99% trace metals basis
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Potassium fluoride, anhydrous, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Phosphorus pentoxide, powder, ACS reagent, ≥98.0%