Merck
  • Associations between Environmental Exposures and Incident Colorectal Cancer by ESR2 Protein Expression Level in a Population-Based Cohort of Older Women.
Associations between Environmental Exposures and Incident Colorectal Cancer by ESR2 Protein Expression Level in a Population-Based Cohort of Older Women.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology (2015-02-05)
Lori S Tillmans, Robert A Vierkant, Alice H Wang, Niloy Jewel Samadder, Charles F Lynch, Kristin E Anderson, Amy J French, Robert W Haile, Lisa J Harnack, John D Potter, Susan L Slager, Thomas C Smyrk, Stephen N Thibodeau, James R Cerhan, Paul J Limburg
ABSTRACT
Cigarette smoking (smoking), hormone therapy (MHT), and folate intake (folate) are each thought to influence colorectal cancer risk, but the underlying molecular mechanisms remain incompletely defined. Expression of estrogen receptor β (ESR2) has been associated with colorectal cancer stage and survival. In this prospective cohort study, we examined smoking, MHT, and folate-associated colorectal cancer risks by ESR2 protein expression level among participants in the Iowa Women's Health Study (IWHS). Self-reported exposure variables were assessed at baseline. Archived, paraffin-embedded colorectal cancer tissue specimens were collected and evaluated for ESR2 protein expression by IHC. Multivariate Cox regression models were fit to estimate relative risks (RR) and 95% confidence intervals (CI) for associations between smoking, MHT, or folate and ESR2-defined colorectal cancer subtypes. Informative environmental exposure and protein expression data were available for 491 incident colorectal cancer cases. Positive associations between ESR2-low and -high tumors and several smoking-related variables were noted, most prominently with average number of cigarettes per day (RR, 4.24; 95% CI, 1.81-9.91 for ESR2-low and RR, 2.15; 95% CI, 1.05-4.41 for ESR2-high for ≥40 cigarettes compared with nonsmokers). For MHT, a statistically significant association with ESR2-low tumors was observed with longer duration of exposure (RR, 0.54; 95% CI, 0.26-1.13 for >5 years compared with never use). No associations were found for folate. In this study, smoking and MHT were associated with ESR2 expression patterns. These data support possible heterogeneous effects from smoking and MHT on ERβ-related pathways of colorectal carcinogenesis in older women.
Product Description
Supelco
Alpha Tocopherol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(±)-α-Tocopherol, tested according to Ph. Eur.
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
USP
Alpha Tocopherol, United States Pharmacopeia (USP) Reference Standard
Supelco
(±)-α-Tocopherol, analytical standard
Sigma-Aldrich
DL-Methionine, 99%, FCC, FG
Sigma-Aldrich
α-Tocopherol, ≥95.5%
Sigma-Aldrich
DL-Methionine, ≥99.0% (NT)
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
DL-Methionine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
DL-Methionine, ≥99%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
α-Tocopherol, European Pharmacopoeia (EP) Reference Standard
Sucrose, European Pharmacopoeia (EP) Reference Standard
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Methionine, European Pharmacopoeia (EP) Reference Standard
DL-Methionine, European Pharmacopoeia (EP) Reference Standard