Merck
  • Combined experimental and in silico approaches for exploring antiperoxidative potential of structurally diverse classes of antioxidants on docetaxel-induced lipid peroxidation using 4-HNE as the model marker.
Combined experimental and in silico approaches for exploring antiperoxidative potential of structurally diverse classes of antioxidants on docetaxel-induced lipid peroxidation using 4-HNE as the model marker.
Bioorganic chemistry (2014-06-03)
Partha Pratim Roy, Sarbani Dey Ray, Supratim Ray
ABSTRACT
The objective of the present work was tantamount to explain the antiperoxidative potential and structural requirements of twenty-eight structurally diverse classes of antioxidants on docetaxel-induced lipid peroxidation. Both experimental and computational approaches were taken to the work. The experiments were performed in vitro and goat liver was used as a source of lipid. 4-hydroxy-2-nonenal was used as model marker for estimation of docetaxel-lipid interaction. The computational portion of the work was limited to QSAR analysis of those antioxidants for better understanding of the structural requirements of antioxidants on docetaxel-lipid interaction. The study was done with freely online available 2D descriptors available on PaDEL (open source). Stepwise regression analysis was used as chemometric tool. The experimental study showed the lipid peroxidation induction capacity of docetaxel. It was also noted that all twenty-eight antioxidants had the ability to suppress the lipid peroxidation. But among them butylated hydroxyl toluene showed the highest potential (-20.5%) and flavone showing lowest potential (-0.8%) to suppress the docetaxel-induced lipid peroxidation. The computational study indicates the importance of topology of the whole molecules, topological distances among atoms within a molecule and specific fragment pattern present in a molecule required for inhibition of lipid peroxidation.
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
Guaiacol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Alpha Tocopherol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Naringenin, natural (US), 98%
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Supelco
Butylated Hydroxytoluene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Kaempferol, analytical standard
Supelco
Apigenin, analytical standard
Sigma-Aldrich
L-Ascorbic acid, 99%
USP
Butylated hydroxytoluene, United States Pharmacopeia (USP) Reference Standard
Supelco
(±)-Naringenin, analytical standard
Sigma-Aldrich
(±)-α-Tocopherol, tested according to Ph. Eur.
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
Galangin, autophagy inducing flavonoid
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
o-Cresol, ≥99%
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG
Supelco
(±)-α-Tocopherol, analytical standard
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
Flavone, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder